Mangan(II)chlorid 1 M in aqueous solution
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
Synonymer: Manganese dichloride
J63150.AD J63150.AE
J63150.ADEA 720 DKK
J63150.AD J63150.AE
Mangan(II)chlorid 1 M in aqueous solution
Manganese(II) chloride
CAS No.: 7773-01-5 Molecular Formula: Cl2Mn Formula Weight: 125.84 Physical Form: Liquid Appearance: Colourless
Formel: MnCl₂
Molekylvægt: 125.84 g/mol
Kogepunkt: 1190
Smeltepunkt: 650
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011114
CAS-nummer: 7773-01-5
EINECS: 231-869-6
Merck Index: 12,05768