3-Fluor-4-nitroanilin ≥95.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
F0787-5G
F0787-1G
TCIAF0787-5GEA
1090
DKK
TCIAF0787-5G
TCIAF0787-1G
3-Fluor-4-nitroanilin ≥95.0% (by GC)
3-Fluor-4-nitroanilin
Formel:
C₆H₅FN₂O₂ Molekylvægt: 156.12 g/mol |
MDL Number:
MFCD03412239 CAS-nummer: 2369-13-3 EINECS: 219-130-6 |
|
Specifikationer Testresultater
Appearance | Light yellow to Brown powder to crystal |
Purity (GC) | min. 95.0 % |
Melting point | 159.0 to 163.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...