Bis(4-Chlorphenyl)sulfon ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
4,4'-Dichloro diphenyl sulphone, 4-Chlorophenyl sulphone, 4,4'-sulphonylbis(chlorobenzene)
B0810-25G
B0810-500G
B0810-100G
TCIAB0810-25GEA
132
DKK
TCIAB0810-25G
TCIAB0810-500G
TCIAB0810-100G
Bis(4-Chlorphenyl)sulfon ≥98.0% (by GC)
Bis(4-Chlorphenyl)sulfon
Formel:
C₁₂H₈Cl₂O₂S |
MDL Number:
MFCD00000619 CAS-nummer: 80-07-9 EINECS: 201-247-9 |
|
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 148.0 to 151.0 deg-C |
Solubility in hot Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...