2'-Chlor-4'-Fluoracetophenon ≥97.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
1-(2-Chloro-4-fluorophenyl)ethanone
C2643-5G
C2643-1G
TCIAC2643-5GEA
739
DKK
TCIAC2643-5G
TCIAC2643-1G
2'-Chlor-4'-Fluoracetophenon ≥97.0% (by GC)
2'-Chlor-4'-Fluoracetophenon
Formel:
C₈H₆ClFO Molekylvægt: 172.59 g/mol Kogepunkt: 110°C |
MDL Number:
MFCD00042202 CAS-nummer: 700-35-6 |
|
Specifikationer Testresultater
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...