Magnesiumchlorid 1 M in aqueous solution
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
J62411.AD J62411.AE
J62411.ADEA 744 DKK
J62411.AD J62411.AE
Magnesiumchlorid 1 M in aqueous solution
Magnesium chloride
CAS No.: 7786-30-3 Molecular Formula: Cl2Mg Formula Weight: 95.22 Physical Form: Liquid MDL No.: MFCD00011106
Formel: MgCl₂
Molekylvægt: 95.21 g/mol
Kogepunkt: 1412
Smeltepunkt: 715
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011106
CAS-nummer: 7786-30-3
EINECS: 232-094-6
Merck Index: 13,05684