N,N,N',N'-Tetramethylethylendiamin for electrophoresis
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Farlig
Synonymer: 1,2-Bis(dimethylamino)ethane, TMEDA, N,N,N',N'-Tetramethylethylenediamine
J63734.AC J63734.AE
J63734.ACEA 580 DKK
J63734.AC J63734.AE
N,N,N',N'-Tetramethylethylendiamin for electrophoresis
N,N,N',N'-Tetramethylethylendiamin
Formel: C₆H₁₆N₂
Kogepunkt: 120…122 °C
Smeltepunkt: –55 °C
Massefylde: 0,775
Flammepunkt: 18 °C (154 °F)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00008335
CAS-nummer: 110-18-9
EINECS: 203-744-6
UN nummer: 2372
ADR: 3,II
Merck Index: 14,9134
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6838f1f5b7d0f69dcc81a8ed247f6c178dad231c&vid=ea361367e9944e4101e38622fdd18b07756eb676&prid=web&clid=SAPDAM