L(+)-Histidin, HCl monohydrat cell culture reagent
Leverandør: Thermo Fisher ScientificSamlet vurdering
Gennemsntilig vurdering
Synonymer:
L-Histidine monohydrochloride monohydrate, (2S)-(+)-Histidine monohydrochloride monohydrate, L-Histidine hydrochloride monohydrate, (S)-(+)-Histidine monohydrochloride monohydrate, His HCl monohydrate, H-His-OH.HCl.H2O, H-His-OH·HCl·H₂O, L-3-Imidazol-4-ylalanine monohydrochloride monohydrate, L-Histidine hydrochloride hydrate
CAS No.: 5934-29-2
Molecular Formula: C6H9N3O2.HCl.H2O
Formula Weight: 209.63 (191.62anhy)
Physical Form: Powder
Appearance: White
Melting Point: ca 240° dec.
Beilstein Reference: 4168261
Merck Reference: 14,4720
Optical Rotation: +10° (c=5 in 5N HCl)
MDL No.: MFCD00151027
Formel:
C₆H₉N₃O₂·HCl·H₂O Smeltepunkt: ∼ 240 °C (dec.) Massefylde: 1,485 g/cm³ (20 °C) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00151027 CAS-nummer: 5934-29-2 EINECS: 211-438-9 Merck Index: 14,4720 |