Tris-(hydroxymethyl) aminomethan (TRIS) 99.9% til elektroforese
Leverandør: Thermo Fisher ScientificSamlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Purity: 99.9%
CAS No.: 77-86-1
Molecular Formula: C4H11NO3
Formula Weight: 121.14
Physical Form: Crystalline powder
Density: 1.353
Appearance: White
Melting Point: 168-172°
Boiling Point: 219-220°/10mm
Beilstein Reference: 741883
Merck Reference: 14,9772
MDL No.: MFCD00004679
Formel:
NH₂C(CH₂OH)₃ Kogepunkt: 219…220 °C (10 mmHg) Smeltepunkt: 168…172 °C Massefylde: 1,353 Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00004679 CAS-nummer: 77-86-1 EINECS: 201-064-4 Merck Index: 14,9772 |