4-(Trifluoromethylthio)benzylamine ≥97.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

T2763-5G T2763-1G
TCIAT2763-5GEA 3620 DKK
TCIAT2763-5G TCIAT2763-1G
4-(Trifluoromethylthio)benzylamine ≥97.0% (by GC, titration analysis)
4-(Trifluoromethylthio)benzylamine
Formel: C₈H₈F₃NS
Flammepunkt: 79°C
MDL Number: MFCD00190129
CAS-nummer: 128273-56-3
UN nummer: 2735
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2771fa065a7f144fcc5932d85f2b5aab1d454378&vid=e0743533ceadb91da6da41a7e93023ae446d2dde&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR