7-Fluoroquinaldine ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

F0519-1G F0519-5G
TCIAF0519-1GEA 600 DKK
TCIAF0519-1G TCIAF0519-5G
7-Fluoroquinaldine ≥98.0% (by titrimetric analysis)
7-Fluoroquinaldine
Formel: C₁₀H₈FN
Kogepunkt: 120°C
MDL Number: MFCD02683029
CAS-nummer: 1128-74-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4a2664d580def7aae498d359d74a3f000f4ddd0a&vid=4a2664d580def7aae498d359d74a3f000f4ddd0a&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR