α,3-Dibromo-4-fluorotoluene ≥97.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 3-Bromo-4-fluorobenzyl bromide, 2-Bromo-4-(bromomethyl)-1-fluorobenzene

B2975-25G B2975-5G
TCIAB2975-25GEA 1200 DKK
TCIAB2975-25G TCIAB2975-5G
α,3-Dibromo-4-fluorotoluene ≥97.0% (by GC)
α,3-Dibromo-4-fluorotoluene
Formel: C₇H₅Br₂F
Molekylvægt: 267.92 g/mol
MDL Number: MFCD09037367
CAS-nummer: 78239-71-1
UN nummer: 3261
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=49cbcc71c562720f7d9ba77459e56af02dc5667b&vid=49cbcc71c562720f7d9ba77459e56af02dc5667b&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow to Light red powder to crystal
Purity (GC) min. 97.0 %
Melting point 39.0 to 43.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR