2-Hydroxyethyl phenyl sulphide ≥97.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
P0733-25ML
P0733-500ML
TCIAP0733-25MLEA
255
DKK
TCIAP0733-25ML
TCIAP0733-500ML
2-Hydroxyethyl phenyl sulphide ≥97.0% (by GC)
2-Hydroxyethyl phenyl sulphide
Formel:
C₈H₁₀OS |
MDL Number:
MFCD00002905 CAS-nummer: 699-12-7 EINECS: 211-828-9 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...