2-Chloroethyl phenyl sulphide ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

C1388-25G C1388-5G
TCIAC1388-25GEA 911 DKK
TCIAC1388-25G TCIAC1388-5G
2-Chloroethyl phenyl sulphide ≥98.0% (by GC)
2-Chloroethyl phenyl sulphide
Formel: C₈H₉ClS
Molekylvægt: 172.68 g/mol
Kogepunkt: 245°C
Flammepunkt: 104°C
MDL Number: MFCD00013691
CAS-nummer: 5535-49-9
EINECS: 226-891-8
UN nummer: 1760
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=60fe3f9b77974c188c28b19f7a838e42b6ff6759&vid=60fe3f9b77974c188c28b19f7a838e42b6ff6759&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR