4-Isobutylphenylboronic acid ≥98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 4-Isobutylbenzeneboronic acid

H52603.03 H52603.06
H52603.03EA 402 DKK
H52603.03 H52603.06
4-Isobutylphenylboronic acid ≥98%
4-Isobutylphenylboronic acid

Purity: 98% CAS No.: 153624-38-5 Molecular Formula: C10H15BO2 Formula Weight: 178.04 Melting Point: 117-120° MDL No.: MFCD01632198

Formel: C₁₀H₁₅BO₂
Smeltepunkt: 117…120 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD01632198
CAS-nummer: 153624-38-5

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR