Sodium-1-tridecanesulphonate ≥98.0% (by titrimetric analysis) til ion-par kromatografi
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
I0351-25G
I0351-5G
TCIAI0351-25GEA
3010
DKK
TCIAI0351-25G
TCIAI0351-5G
Sodium-1-tridecanesulphonate ≥98.0% (by titrimetric analysis) til ion-par kromatografi
Sodium-1-tridecanesulphonate
Formel:
C₁₃H₂₇NaO₃S |
MDL Number:
MFCD00024993 CAS-nummer: 5802-89-1 |
|
Specifikationer Testresultater
Appearance | White powder to crystal |
Purity (Ion exchange titration) | min. 98.0 % |
Absorbance | to pass test |
Solubility in hot Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...