Sodium-1-hexadecanesulphonate

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Sodium hexadecane-1-sulphonate, Hexadecane-1-sulphonic acid sodium salt

H0415-10G H0415-25G
TCIAH0415-10GEA 444 DKK
TCIAH0415-10G TCIAH0415-25G
Sodium-1-hexadecanesulphonate
Sodium-1-hexadecanesulphonate
Formel: C₁₆H₃₃NaSO₃
Molekylvægt: 328.49 g/mol
MDL Number: MFCD00007528
CAS-nummer: 15015-81-3
EINECS: 239-100-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=63cc5535bb7b50e1f0f3fc443f57cfc168a7ecb9&vid=63cc5535bb7b50e1f0f3fc443f57cfc168a7ecb9&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Solubility in hot Water almost transparency
Drying loss max. 2.0 %
Ignition residue (Sulfate) 20.8 to 21.9 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR