4-Chloro-2,6-xylidine ≥98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 4-Chloro-2,6-dimethylaniline

B21996.06 B21996.14
B21996.06EA 508 DKK
B21996.06 B21996.14
4-Chloro-2,6-xylidine ≥98%
4-Chloro-2,6-xylidine

Purity: 98% CAS No.: 24596-18-7 Molecular Formula: C8H10ClN Formula Weight: 155.63 Melting Point: 45-50° MDL No.: MFCD01076576

Formel: C₈H₁₀ClN
Smeltepunkt: 45…50 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD01076576
CAS-nummer: 24596-18-7

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR