p-Toluenesulphonyl fluoride ≥98%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
p-toluenesulphonylfluoride
L12204.06
L12204.14
L12204.06EA
406
DKK
L12204.06
L12204.14
p-Toluenesulphonyl fluoride ≥98%
p-Toluenesulphonyl fluoride
Purity: 98%
CAS No.: 455-16-3
Molecular Formula: C7H7FO2S
Formula Weight: 174.19
Flash Point: 105°(221°F)
Melting Point: 40-41°
Boiling Point: 112°/16mm
Beilstein Reference: 2208423
UN No.: 3261
MDL No.: MFCD00007421
Formel:
C₇H₇FO₂S Kogepunkt: 112 °C (16 mmHg) Smeltepunkt: 40…41 °C Flammepunkt: 105 °C (221 °F) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00007421 CAS-nummer: 455-16-3 EINECS: 207-238-6 UN nummer: 3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...