4-Nitrophenyl phenyl sulphide ≥98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: (4-Nitrophenyl)(phenyl)sulphane

A17278.06 A17278.14
A17278.06EA 778 DKK
A17278.06 A17278.14
4-Nitrophenyl phenyl sulphide ≥98%
4-Nitrophenyl phenyl sulphide

Purity: 98% CAS No.: 952-97-6 Molecular Formula: C12H9NO2S Formula Weight: 231.27 Melting Point: 54-57° MDL No.: MFCD00024700

Formel: C₁₂H₉NO₂S
Molekylvægt: 231.27 g/mol
Smeltepunkt: 54…57 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00024700
CAS-nummer: 952-97-6
EINECS: 213-462-5

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR