4-(Ethoxycarbonyl)phenylboronic acid (contains varying amounts of Anhydride)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

E0868-1G E0868-5G
TCIAE0868-1GEA 321 DKK
TCIAE0868-1G TCIAE0868-5G
4-(Ethoxycarbonyl)phenylboronic acid (contains varying amounts of Anhydride)
4-(Ethoxycarbonyl)phenylboronic acid
Formel: C₉H₁₁BO₄
Smeltepunkt: 160°C
MDL Number: MFCD02179441
CAS-nummer: 4334-88-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=08d7ee063fed3e6148eb2455a3861a295d2bbf4a&vid=08d7ee063fed3e6148eb2455a3861a295d2bbf4a&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 98.0 to 111.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR