2-(Trifluoromethyl)acrylic acid 98%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
2-(Trifluoromethyl)propenoic acid
313100010
313100050
ACRO313100010EA
325
DKK
ACRO313100010
ACRO313100050
2-(Trifluoromethyl)acrylic acid 98%
2-(Trifluoromethyl)acrylic acid
Formel:
C₄H₃F₃O₂ Molekylvægt: 140.06 g/mol Kogepunkt: 86 °C (760 mmHg) Smeltepunkt: 48…52 °C Flammepunkt: 81 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00042424 CAS-nummer: 381-98-6 |
Specifikationer Testresultater
Appearance | Transparent light yellow Chunks |
Infrared spectrum | Conforms |
Melting point | 48 to 52 °C |
Titration with TBAH | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...