Divinyl sulphone ≥96.0% (by GC) stabilised
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Stabilization:
Stabilised with hydroquinone (HQ)
D0959-25G
D0959-250G
D0959-5G
TCIAD0959-25GEA
411
DKK
TCIAD0959-25G
TCIAD0959-250G
TCIAD0959-5G
Divinyl sulphone ≥96.0% (by GC) stabilised
Divinyl sulphone
Formel:
C₄H₆O₂S Molekylvægt: 118.16 g/mol Kogepunkt: 110°C Flammepunkt: 102°C Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD00008623 CAS-nummer: 77-77-0 EINECS: 201-057-6 UN nummer: 2927 ADR: 6.1 I |
|
Specifikationer Testresultater
Appearance | Colorless to Red to Green clear liquid |
Purity (GC) | min. 96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...