N,N,N',N'-Tetrakis(2-hydroxypropyl)ethylenediamine ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

T0781-25G T0781-500G
TCIAT0781-25GEA 140 DKK
TCIAT0781-25G TCIAT0781-500G
N,N,N',N'-Tetrakis(2-hydroxypropyl)ethylenediamine ≥98.0% (by titrimetric analysis)
N,N,N',N'-Tetrakis(2-hydroxypropyl)ethylenediamine
Formel: C₁₄H₃₂N₂O₄
Flammepunkt: 211°C
MDL Number: MFCD00004534
CAS-nummer: 102-60-3
EINECS: 203-041-4
Merck Index: 14,03599
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=16fd727512b8aaea7538075a020b14f348cfd675&vid=16fd727512b8aaea7538075a020b14f348cfd675&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR