Manual crimping tools, standard version
Leverandør: MACHEREY-NAGEL
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
735111 735113 735120 735126 735408 735911 735913 735920 735132 735133
MANA735111EA 3440 DKK
MANA735111 MANA735113 MANA735120 MANA735126 MANA735408 MANA735911 MANA735913 MANA735920 MANA735132 MANA735133
Manual crimping tools, standard version
Tænger til Vials
These manual crimping tools are made of robust metal.
Manual crimpers
• Adjustable crimping height via hexagon key (not for 8 mm)
• Crimping pressure adjustable via screw in the handle
• Also for Flip Top / Flip Off caps available
Manual decappers
• Allow safe removal of caps
• No adjustment required
• Easy to handle decapper in plier style for 11 mm and 20 mm caps is avaialble