Laminated steel padlocks
Leverandør: Brady
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
814104 814086 814087 814089 814090 814091 814092 814093 814094 814095 814096 814088 814099 814097 814112 814100 814101 814102 814103 814105 814106 814107 814108 814109 814111 814098 814110
238-0072EA 1380 DKK
238-0072 238-0054 238-0055 238-0057 238-0058 238-0059 238-0060 238-0061 238-0062 238-0063 238-0064 238-0056 238-0067 238-0065 238-0080 238-0068 238-0069 238-0070 238-0071 238-0073 238-0074 238-0075 238-0076 238-0077 238-0079 238-0066 238-0078
Laminated steel padlocks
Sikkerhedslåse
These robust padlocks withstand severe physical abuse and come with a hardened steel shackle making them extremely difficult to cut. They are ideal for use outdoors and in high temperature environments (max. 580 °C).
  • Reinforced laminated steel, with steel shackle
  • Body plates are individually coated for superior rust protection
  • 5-pin cylinder offers more unique key cuts and better tamper resistance than conventional 4-pin locks
  • Coloured bumpers can be used for identification
  • Key will not be released until the padlock shackle is closed
  • Each padlock is keyed differently and is supplied with one key
Shackle diameter: 6,5 mm