PIG® eco-friendly spill response bag
Leverandør: NEW PIG
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
KITE211 RFLE211
121-0065EA 653 DKK
121-0065 121-0121
PIG® eco-friendly spill response bag
Spild materialer
This compact "Green" bag makes transport simple, and its recycled-content pads and socks soak up more liquid with less environmental impact.
  • Environmentally friendly absorbents feature high recycled content; reuses material that would normally enter a waste stream
  • Reusable spill bag is constructed out of jute plant bark; naturally strong and durable with waterproof inner coating to keep kit clean, dry and ready to use
  • High-visibility colour helps identify spill kit location; saves valuable time during response
  • Hook-and-loop closure is easy to open for fast access to absorbents
  • Carry handle affords convenient transport; makes it ideal for hanging on a wall, storing in a cabinet or being carried in a vehicle cab
  • Compact design takes up little storage room and effectively deals with minor spillages
The super-absorbent PIG BLUE® mat pads and PIG® SUPER socks have maximum capacity for containing and soaking up spills of oils, coolants, solvents or water.
Pads: Natural and recycled fibres
Socks: Recycled cellulose filler, polypropylene skin
Disposal bags: Regran polyethylene (recycled polymers)
Spill kit bag: Jute