T-shirts, Turin B195
Leverandør: Portwest
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
B195WHRXXL B195WHRXS B195WHRXXXL B195WHRXL B195WHRS B195WHRM B195WHRL B195NARXXXL B195NARXXL B195NARXL B195NARS B195NARL B195NARM B195HGRL B195HGRS B195HGRXL B195HGRXXXL B195HGRM B195HGRXXL
113-6212EA 110 DKK
113-6212 113-6896 113-6213 113-6211 113-6208 113-6209 113-6210 113-6219 113-6218 113-6217 113-6214 113-6216 113-6215 PRTWB195HGRL PRTWB195HGRS PRTWB195HGRXL PRTWB195HGRXXXL PRTWB195HGRM PRTWB195HGRXXL
T-shirts, Turin B195
Arbejdstøj
These high quality shirts are made of 100% cotton.
  • Round neck
  • Taped neckline
Weight: 195 g/m²