CHROMABOND® Vacuum Manifolds
Leverandør: MACHEREY-NAGEL
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
730150N 730151N
MANA730150NEA 13830 DKK
MANA730150N MANA730151N
CHROMABOND® Vacuum Manifolds
Manifolder
CHROMABOND® Vacuum manifolds are designed for reproducible SPE applications.
  • For simultaneous preparation of up to 12, 16 or 24 samples
  • Available replacement parts for vacuum manifolds and accessories for special applications
Leveringsinformation: The vacuum manifold complete consists of glass cabinet with lid and lid gasket, removable needles on lower side of lid, vacuum gauge, control valve, valves and caps, variable rack, and PP tank (only for MANA730150).