Nitrile gloves, Showa 8005PF
Leverandør: Showa
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
8005PFL 8005PFM 8005PFS 8005PFXL
112-2867EA 316 DKK
112-2867 112-2868 112-2869 112-2870
Nitrile gloves, Showa 8005PF
Handsker
The SHOWA 8005PF disposable glove is ideal for jobs requiring increased protection without compromising on tactility.
  • Rolled cuff prevents dirt from entering the glove and allows for easy removal of the glove
  • Second-skin feel to significantly reduce hand fatigue over long periods of wear
  • Silicone- and powder-free
  • Latex-free
  • Ambidextrous
By using SHOWA’s Eco Best Technology®, the 8005PF’s are completely eco-friendly, breaking down by 82% in just 386 days when tested in a laboratory.
EN 374 (JKL), EN 388 (1000); Cat. III; AQL 1,5
Certificeringer: EN ISO 374-1 Type B (JKOPT), EN ISO 374-5 VIRUS; Cat. III
Food contact approved.