SPE columns, HyperSep™ silica
Leverandør: Thermo Fisher Scientific
Ratings:
Gennemsntilig vurdering: 5.0 (Bedømmelse)
Samlet vurdering
Gennemsntilig vurdering
Var denne bedømmelse brugbar? Ja Nej
Ja: 0 Nej: 0 
Chimie analytique / Chimiste / LYON
Ja, jeg anbefaler
Très adapté pour faire de la purification d'échantillon liquide à matrice complexe.
60108-711 60108-410 60108-426 60108-411 60108-317 60108-315 60108-409 60108-710 60108-712 60108-851 60108-853 60108-793
549-0894EA 1600 DKK
549-0894 549-0890 549-0892 549-0891 549-0888 549-0887 549-0889 549-0893 549-0895 549-0898 549-0956 549-0896
SPE columns, HyperSep™ silica
Kromatografikolonner
A polar sorbent primarily used to retain analytes in non-polar matrices.
  • Effectively separates compounds of very similar structure
  • Adsorbs analytes from non-polar solvents like hydrocarbons, less polar esters and ethers
  • Suitable for use as an intermediate strength cation exchanger in aqueous medium