Cut resistant gloves, HyFlex® 11-630 / 11-638
Leverandør: Ansell
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
11-630/8 11-638/10 11-638/8 11-638/9
112-2924EA 0 DKK
112-2924 112-3148 112-3152 112-3153
Cut resistant gloves, HyFlex® 11-630 / 11-638
Handsker
The gloves are designed for use in dry, as well as slightly oily environments. The nylon/Lycra®/Kevlar®/cotton liners have a yarn structure that contains an optimal ratio of HPPE (High Performance PE), providing high levels of cut resistance.
  • Palm coated with polyurethane for additional protection and abrasion resistance
  • Specific formulation of the polyurethane dipping provides a secure grip on dry or slightly oily pieces
  • Lycra® liner and PU coating provide excellent flexibility and fit
  • Elasticated cuffs
Ideal for plastics injection and moulding, assembling metal parts and components and the handling of sharp or irregular-edged metal parts.
Colour: Grey
EN 388 (4443), Cat. II
Certificeringer: EN 388:2016 (4443B), Cat. II