Capillary columns, CP-Wax 57 CB
Leverandør: VARIAN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
CP7615 CP7617
VARICP7615EA 10770 DKK
VARICP7615 VARICP7617
Capillary columns, CP-Wax 57 CB
Kromatografikolonner
CP-Wax 57 CB for Glycols and Alcohols is guaranteed for the analysis of glycols, diols and alcohols. It has a unique, high polarity wax phase that produces symmetrical peaks.
  • Guaranteed analysis for complete confidence
  • Symmetrical peaks providing the most accurate results
  • Extensive cross-linking delivers robustness and enhanced column lifetime