Screw neck vials, N 10
Leverandør: MACHEREY-NAGEL
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
702013 702011 702012
MANA702013EA 251 DKK
MANA702013 MANA702011 MANA702012
Screw neck vials, N 10
Hætteglas
These screw neck vials have a wide opening for easy filling.
  • Made of clear and amber glass
  • Flat bottom
  • Label and scale for convenient handling
  • Silanised inserts for highest demands are available
Often used on PerkinElmer, Shimadzu and Waters instruments.
Screw closures are bonded (septa firmly connected with the cap) or ready assembled.
- Large range of bonded screw caps with different septum materials
- Septa made from economical Red Rubber/TEF or high quality silicone/PTFE (also slitted)