Partikel filtrerings halvmasker, til engangsbrug, FFP1/FFP2/FFP3, X-plore® 1300
Leverandør: Dräger
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
3951215 3951217 3951211 3951214 3951216 3951213 3951218 3951212
111-1040EA 705 DKK
111-1040 111-1044 111-0759 111-0760 111-1043 111-0907 111-1045 111-1545
Partikel filtrerings halvmasker, til engangsbrug, FFP1/FFP2/FFP3, X-plore® 1300
Åndedrætsværn Engangsmasker Partikelmasker
The X-plore® 1300 series offers models in all three EN protection classes: FFP1, FFP2 and FFP3. Available with or without CoolMax™ exhalation valve for reduced exhalation resistance and to avoid a build-up of heat. Odour models for tackling unpleasant odours are equipped with an additional activated carbon layer.
  • Special high performance filter media are brought together to form the CoolSafe™ filter material, ensuring a low level of exhalation resistance for users while delivering a high level of filter performance
  • Flexible nose clip ensures the product is easy to adjust to the face
  • Sweat-absorbing sealed padding to reliably seal off the nasal area
  • Ergonomically shaped, solid mask bodies offer a comfortable and secure fit for a wide range of facial shapes
  • VarioFlex™ head strap with EasyStop mechanism to make it easy to put on and take off - easy to adjust and provides a pressure-free fit
  • Inner fleece is soft on the skin and ensures the product is comfortable to wear
  • The colour coding system on the nose clip and valve ensures greater levels of safety, with dark blue, light blue and white used to indicate the three protection levels
Certificeringer: EN 149:2001+A1:2009; tested according to the dolomite dust test.
Emballering: Each mask is individually packed in a plastic pouch.
Advarsel: *One size fits all