Snap ring caps for snap ring vials, N 11
Leverandør: MACHEREY-NAGEL
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
702710 702718 702064 702063 702063.2080 702095 702108 702142 702143 702710.1 702710.2080 702717.2 702717.2080 702718.1 702718.2080 702731
MANA702710EA 337 DKK
MANA702710 MANA702718 MANA702064 MANA702063 MANA702063.2080 MANA702095 MANA702108 MANA702142 MANA702143 MANA702710.1 MANA702710.2080 MANA702717.2 MANA702717.2080 MANA702718.1 MANA702718.2080 MANA702731
Snap ring caps for snap ring vials, N 11
Låg Låg til kromatografirør
Ready assembled snap ring closures made of polyethylene are available for the snap ring vials.
  • Available as hard cap version in transparent and further colours or as soft cap in light blue for easy handling
  • Large choice of septa materials
  • Most frequent version are closures with cross-slit silicone/PTFE septum, which supports easy penetration of blunt HPLC needles