Volumetric flasks, class A, white markings, USP batch certificate, BLAUBRAND®
Leverandør: Brand
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
37462 37465 37464 37467 37468 37469 37470 37471 37461 37482 37483 37484 37485 37487 37488 37489 37463 37481 37490 37491
612-5420EA 884 DKK
612-5420 612-5423 612-5422 612-5424 612-5425 612-5426 612-5427 612-5428 612-5419 612-4825 612-4826 612-4827 612-4828 612-4829 612-4830 612-4831 612-5421 612-4824 612-4832 612-4833
Volumetric flasks, class A, white markings, USP batch certificate, BLAUBRAND®
Kolber Volumetriske kolber
Borosilicate glass 3.3, amber, with amber glass stopper or PP stopper.
  • Calibrated to contain (TC, In)
  • Marks and inscriptions in high contrast white enamel
  • DE-M marking
  • With USP batch certificate, on request also available with USP individual certificate
DIN EN ISO 1042
Certificeringer: DIN EN ISO 1042.
Bestil nu
Specifikationer
Environmentally Preferable Bæredygtig emballage Bæredygtig emballage Bæredygtig emballage Bæredygtig emballage Bæredygtig emballage Bæredygtig emballage Bæredygtig emballage Bæredygtig emballage Bæredygtig emballage Bæredygtig emballage