Safety shoes, slip-on, Jalbio
Leverandør: Jallatte
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
J0271/35 J0271/36 J0271/37 J0271/38 J0271/39 J0271/40 J0271/41 J0271/42 J0271/43 J0271/44 J0271/45 J0271/46 J0271/47
JALCJ0271/35EA 1330 DKK
JALCJ0271/35 JALCJ0271/36 JALCJ0271/37 JALCJ0271/38 JALCJ0271/39 JALCJ0271/40 JALCJ0271/41 JALCJ0271/42 JALCJ0271/43 JALCJ0271/44 JALCJ0271/45 JALCJ0271/46 JALCJ0271/47
Safety shoes, slip-on, Jalbio
Sko og støvler
Slip-on safety shoes made from Aseptane™ upper (resistant to fluid projection) and Softane™/Néotril™ sole (heat resistant up to 300 °C).
  • Aseptane™ upper is water repellent, breathable, washable at 30°C
  • Anatomically shaped Confort-Tech insole; anti-static and antibacterial, with shock absorber heel
  • Lining made from anti-abrasion and breathable fibre
  • Slip resistant Triftane™ sole helps to prevent fatigue
  • Perforated steel toecap (200 J)
Suitable for food industries and wet working conditions.
Colour: White
Certificeringer: EN 345-1; S2, HRO