Capillary columns, CP-Sil 5/C₁₈ CB for PCB
Leverandør: VARIAN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
CP7476 CP7477
VARICP7476EA 35410 DKK
VARICP7476 VARICP7477
Capillary columns, CP-Sil 5/C₁₈ CB for PCB
Kromatografikolonner
CP-Sil 5/C₁₈ CB for PCB has a lower polarity than 100% polydimethylsiloxane due to its C₁₈ substitutions. Due to the absence of cyano groups it provides high signal-to-noise ratios for ECD detectors.
  • Guaranteed for very high resolution PCB analysis for ultimate confidence
  • 100 m column separates critical isomer pairs for accurate results
  • Use with high sensitivity ECD detection for enhanced productivity