Crimp Caps for Crimp Neck Vials, N 11
Leverandør: MACHEREY-NAGEL
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
70231 70231.1 70231.2 70239 702401 70256 702801 70284 70288 702995 702823 702730 702175 702730.1 702730.2 702730.3 702879 70288.1 70288.2 70288.3 702995.1 702995.2 702995.3 70246.3 70246
MANA70231EA 261 DKK
MANA70231 MANA70231.1 MANA70231.2 MANA70239 MANA702401 MANA70256 MANA702801 MANA70284 MANA70288 MANA702995 MANA702823 MANA702730 MANA702175 MANA702730.1 MANA702730.2 MANA702730.3 MANA702879 MANA70288.1 MANA70288.2 MANA70288.3 MANA702995.1 MANA702995.2 MANA702995.3 MANA70246.3 MANA70246
Crimp Caps for Crimp Neck Vials, N 11
Låg Låg til kromatografirør
Ready-to-use crimp caps with assembled septa and plain crimp caps are available for the crimp neck vials.
  • Broad range of aluminium crimp and magnetic crimp closures with different septum materials and cap colours
  • Magnetic version for CTC PAL autosampler or identical constructions
  • Large choice of septa materials