Kompakt dobbelt skovlsystem
Leverandør: CONTEC
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
UBKT0005 UBKT0006
141-0985EA 768 DKK
141-0985 141-0986
Kompakt dobbelt skovlsystem
Mopper
Stainless cart with removable handle and two 5 gallon (20 L) polypropylene buckets. This bucket system features a stainless steel rolling cart with two autoclavable polypropylene buckets, one grey and one red, and options for a heavy-duty stainless steel downpress wringer or flat mop wringer (purchased separately). Compact design improves manoeuverability and reduces storage space. The 2757 cart features a removable handle for easier autoclaving and storage.
  • Ideal for cleanrooms and controlled environments with small autoclaves or limited storage space
  • Compatible with all Contec mops, depending upon wringer choice
The durable yet lightweight cart has four casters, two fixed and two that swivel and lock easily with the touch of a foot. The system can be used with two or three buckets, depending upon the cleaning protocols. An extendable frame pulls out of the front of the cart to accommodate an additional bucket. Included stainless steel connecting kit prevents drips between buckets.