Capillary columns, CP-Sil 88 for FAME
Leverandør: VARIAN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
CP7487 CP7488 CP7489
568-0701EA 19770 DKK
568-0701 568-0702 568-0703
Capillary columns, CP-Sil 88 for FAME
Kromatografikolonner
Guaranteed analysis of FAME cis/trans isomers for complete confidence.
  • High polarity stationary phase providing more efficiency and higher productivity
  • Use for FAME in the C6 to C26 range
Temperature limits: 225/240 °C.