VWR®, Sealing film for ELISA and general incubation
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
731-0309EA 1650 DKK
731-0309 731-0310 731-0311 731-0312
VWR®, Sealing film for ELISA and general incubation
Forsegling Mikrotiterpladetætninger
Polyester film with acrylic adhesive. These non pierceable films in sheeted-roll format are recommended for ELISA tests, general incubation and short-term storage.
  • Recommended temperature range from −40 to +120 °C
  • Total thickness 64,8 μm (film 38,1 μm, adhesive 26,7 μm)
  • Pre-scored films separated by green colour bands for easy application
  • Minimise evaporation and prevent contamination between wells
  • Pre-cut films on a roll
Leveringsinformation: Each roll contains sufficient film to cover 100 microplates.