Industrielle balancer, Ranger® 3000 Count kompakt vægt
Leverandør: OHAUS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
30031786 30031783 30031784 30031782 30060907 30031785 30060911 30060910 30060908 30060909
611-2590EA 4630 DKK
611-2590 611-2587 611-2588 611-2586 611-2612 611-2589 611-2616 611-2615 611-2613 611-2614
Industrielle balancer, Ranger® 3000 Count kompakt vægt
Vægte
A multipurpose, compact counting scale for producing precise results rapidly with weighing, parts counting, check counting and accumulation functions. User friendly design with three dedicated backlit diplays, eight function keys, three check weighing, indicators and full numeric keypad. Stainless steel platform and ABS housing for easy cleaning.
  • Three separate backlit displays for count, weight and piece weight
  • Mains supply with internal rechargeable battery for portable use
  • Internal memory with a 30 item library function for data including average piece weights, check counting or check weighing, parameters, tare weights and accumulated weights or counts
  • Stabilisation within 1 second
Bestillingsoplysninger: More connectivity options available including optional Ethernet, 2nd RS232 or USB and optional 2nd platform. Approved versions are also available. Please see the website or contact your local Avantor sales office for more details.