Capillary columns, CP-Sil PONA CB
Leverandør: VARIAN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
CP7531 CP7530 CP7531I5
568-0705EA 22900 DKK
568-0705 568-0704 568-0706
Capillary columns, CP-Sil PONA CB
Kromatografikolonner
CP-Sil PONA CB delivers accurate analysis of paraffins, olefins, naphthalenes and aromatics in complex hydrocarbon mixtures.
  • Guaranteed hydrocarbon analysis according to ASTM (DHA method)
  • Inert to polar compounds for highly accurate data
  • Excellent reproducibility increases productivity
Temperature limits: 250/275 °C
Similar phases: Petrocol DH, SPB-1, 007-1, Rtx-1, MXT-1