Single channel macro pipettes, electronic, variable volume, Acura® electro 936
Leverandør: Socorex Isba
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
936.10E 936.10 936.02E 936.05 936.02 936.05E 936.02G 936.05G 936.10G
SOCO936.10EEA 6660 DKK
SOCO936.10E SOCO936.10 SOCO936.02E SOCO936.05 SOCO936.02 SOCO936.05E SOCO936.02G SOCO936.05G SOCO936.10G
Single channel macro pipettes, electronic, variable volume, Acura® electro 936
Pipetter Automatiske pipetter
Electronic macro pipette with microprocessor-controlled motor and rechargeable battery pack.
  • Interchangeable nozzle protection filter
  • Outstanding accuracy and precision
  • Ready-to-use packages available
Bestillingsoplysninger: The pipette package is supplied with individual QC certificate and operating instructions, pipette charging stand, fast charger with AC adapter and cord, additional battery pack and Qualitips® pipette tip samples.
The individual pipette is supplied with QC certificate and operating instructions, as well as samples of Qualitips®.