Standard conical ground joints cones, long tip, Quickfit®
Leverandør: SciLabware
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
CBB19UB CBB29UB CBB14UB CBB24UB
201-8201EA 849 DKK
201-8201 201-8202 201-8200 217-0741
Standard conical ground joints cones, long tip, Quickfit®
Adapter Adaptere med NS-slib
PYREX® borosilicate glass.
  • Useful for construction of stirrer guides
  • Avoids the problem of the badge charring at high temperatures which gives an untidy appearance
  • Long tip cones manufactured with a minimum shank
Unbadged ground glass cones are free from ceramic markings.
Total length: 90 mm