2-(2-Bromoethoxy)tetrahydropyran ≥95.0% (by GC) stabilised
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
2-(2-Bromoethoxy)tetrahydro-2H-pyran
Stabilization:
Stabilised with potassium carbonate
B2850-25G
B2850-5G
TCIAB2850-25GEA
1830
DKK
TCIAB2850-25G
TCIAB2850-5G
2-(2-Bromoethoxy)tetrahydropyran ≥95.0% (by GC) stabilised
2-(2-Bromoethoxy)tetrahydropyran
Formel:
C₇H₁₃BrO₂ Kogepunkt: 75°C Flammepunkt: 87°C Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD01321310 CAS-nummer: 17739-45-6 |
|
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...