(2-Hydroxylpropyl)-β-cyclodextrin

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Hydroxypropyl Betadex, Hydroxypropyl-β-cyclodextrin, (2-Hydroxypropyl)-β-cyclodextrin, HP-β-CD

H0979-25G H0979-100G
TCIAH0979-25GEA 476 DKK
TCIAH0979-25G TCIAH0979-100G
(2-Hydroxylpropyl)-β-cyclodextrin
(2-Hydroxylpropyl)-β-cyclodextrin
Formel: C₆₃H₁₁₂O₄₂
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00074978
CAS-nummer: 128446-35-5
EINECS: 420-920-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=42bcb7949075f595c5946d961028d276b97d5d4d&vid=42bcb7949075f595c5946d961028d276b97d5d4d&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Specific rotation [a]20/D +120 to +135 deg (C=1, H2O)
Drying loss max. 10.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR