Kaliumethyl malonat ≥98.0% (by titrimetric analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
Monoethyl malonate potassium salt, Ethyl potassium malonate
M0975-25G
M0975-250G
TCIAM0975-25GEA
247
DKK
TCIAM0975-25G
TCIAM0975-250G
Kaliumethyl malonat ≥98.0% (by titrimetric analysis)
Kaliumethyl malonat
Formel:
C₅H₇KO₄ |
MDL Number:
MFCD00035603 CAS-nummer: 6148-64-7 EINECS: 228-156-7 |
|
Specifikationer Testresultater
Appearance | White powder to crystal |
Purity (Nonaqueous Titration) | 98.0 to 102.0 % |
Solubility in Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...