Aminoacetaldehyddiethylacetal ≥97.0% (by GC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
2,2-Diethoxyethanamine, 2-Aminoacetaldehyde diethylacetal
A0247-25ML
A0247-250ML
TCIAA0247-25MLEA
386
DKK
TCIAA0247-25ML
TCIAA0247-250ML
Aminoacetaldehyddiethylacetal ≥97.0% (by GC, titration analysis)
Aminoacetaldehyddiethylacetal
Formel:
C₆H₁₅NO₂ Molekylvægt: 133.19 g/mol Kogepunkt: 163°C Flammepunkt: 45°C |
MDL Number:
MFCD00008136 CAS-nummer: 645-36-3 EINECS: 211-439-4 UN nummer: 2924 ADR: 3 III |
|
Specifikationer Testresultater
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 97.0 % |
Purity (Nonaqueous Titration) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...